missing translation for 'onlineSavingsMsg'
Learn More

(4-Isopropylphenoxy)acetyl chloride, 98%, Thermo Scientific Chemicals

Catalog No. p-7030342
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2803606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2803606 Supplier Thermo Scientific Chemicals Supplier No. H2803606

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 223128-33-4 | Molecular Formula: C11H13ClO2 | Molecular Weight: 212.673 g/mol

Used as a primary and secondary intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as a primary and secondary intermediate for pharmaceutical.

Solubility
Reacts with water.

Notes
Moisture Sensitive, store away from water/moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under dry inert gas.

Chemical Identifiers

CAS 223128-33-4
Molecular Formula C11H13ClO2
Molecular Weight (g/mol) 212.673
MDL Number MFCD02628421
InChI Key YGVLHWFQKYVLFY-UHFFFAOYSA-N
Synonym 4-isopropylphenoxy acetyl chloride, 2-4-isopropylphenoxy acetyl chloride, acmc-1cokt, 4-iso propyl phenoxy acetyl chloride, 4-isopropyl-phenoxy-acetyl chloride, 4-propan-2-yl phenoxy acetyl chloride, 2-4-methylethyl phenoxy acetyl chloride, 2-4-propan-2-yl phenoxy acetyl chloride, acetyl chloride, 4-1-methylethyl phenoxy, acetyl chloride,2-4-1-methylethyl phenoxy
PubChem CID 21209244
IUPAC Name 2-(4-propan-2-ylphenoxy)acetyl chloride
SMILES CC(C)C1=CC=C(C=C1)OCC(=O)Cl

Specifications

Refractive Index 1.5195
Quantity 5 g
UN Number UN3265
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 212.67
Percent Purity 98%
Chemical Name or Material (4-Isopropylphenoxy)acetyl chloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only