missing translation for 'onlineSavingsMsg'
Learn More

4-Isopropylphenol 99.0+%, TCI America™

Catalog No. p-7131299
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I026225G 25 g
I0262100G 100 g
I0262500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. I026225G Supplier TCI America Supplier No. I026225G
Only null left

Chemical Identifiers

CAS 99-89-8
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.194
MDL Number MFCD00002372
InChI Key YQUQWHNMBPIWGK-UHFFFAOYSA-N
Synonym 4-isopropylphenol, p-isopropylphenol, p-cumenol, australol, phenol, 4-1-methylethyl, 4-1-methylethyl phenol, 1-hydroxy-4-isopropylbenzene, 4-propan-2-yl phenol, phenol, p-isopropyl, para-isopropylphenol
PubChem CID 7465
IUPAC Name 4-propan-2-ylphenol
SMILES CC(C)C1=CC=C(C=C1)O

Specifications

Melting Point 61°C
Boiling Point 109°C
Quantity 25 g
UN Number 2430
Formula Weight 136.19
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Isopropylphenol
EINECSNumber (3)-0503&(4)-0057
RTECSNumber SL5950000
TSCA Yes