missing translation for 'onlineSavingsMsg'
Learn More

4-Isopropylcyclohexylamine (cis- and trans- mixture) 98.0+%, TCI America™

Catalog No. i08535ml
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I08535ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I08535ML Supplier TCI America Supplier No. I08535ML
Only null left

Chemical Identifiers

CAS 52430-81-6
Molecular Formula C9H19N
Molecular Weight (g/mol) 141.258
MDL Number MFCD16853065
InChI Key MFRKYEJMLQUSJX-UHFFFAOYSA-N
Synonym 1-Amino-4-isopropylcyclohexane
PubChem CID 421053
IUPAC Name 4-propan-2-ylcyclohexan-1-amine
SMILES CC(C)C1CCC(CC1)N

Specifications

Color Colorless
Boiling Point 90°C
Quantity 5 mL
UN Number 2733
Formula Weight 141.26
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-Isopropylcyclohexylamine (cis- and trans- mixture)