missing translation for 'onlineSavingsMsg'
Learn More

4-Isopropylcinnamic acid, predominantly trans, 98+%

CAS: 3368-21-6 | C12H14O2 | 190.24 g/mol

$143.34 - $517.23

Chemical Identifiers

CAS 3368-21-6
Molecular Formula C12H14O2
Molecular Weight (g/mol) 190.24
MDL Number MFCD00016544
InChI Key SJDOOXOUSJDYFE-VMPITWQZSA-N
Synonym 4-isopropylcinnamic acid, 3-4-isopropylphenyl acrylic acid, p-isopropylcinnamic acid, 2e-3-4-isopropylphenyl acrylic acid, cinnamic acid, p-isopropyl, 2-propenoic acid, 3-4-1-methylethyl phenyl, 2e-3-4-propan-2-yl phenyl prop-2-enoic acid, e-3-4-isopropylphenyl acrylic acid, e-3-4-isopropyl-phenyl-acrylic acid, 2e-3-4-isopropylphenyl prop-2-enoic acid
PubChem CID 719753
IUPAC Name (E)-3-(4-propan-2-ylphenyl)prop-2-enoic acid
SMILES CC(C)C1=CC=C(\C=C\C(O)=O)C=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1515606
View Documents
Thermo Scientific Chemicals
A1515606
5 g
Each for $143.34
Only null left
 
AAA1515614
View Documents
Thermo Scientific Chemicals
A1515614
25 g
Each for $517.23
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

3368-21-6
190.24
SJDOOXOUSJDYFE-VMPITWQZSA-N
719753
CC(C)C1=CC=C(\C=C\C(O)=O)C=C1
C12H14O2
MFCD00016544
4-isopropylcinnamic acid, 3-4-isopropylphenyl acrylic acid, p-isopropylcinnamic acid, 2e-3-4-isopropylphenyl acrylic acid, cinnamic acid, p-isopropyl, 2-propenoic acid, 3-4-1-methylethyl phenyl, 2e-3-4-propan-2-yl phenyl prop-2-enoic acid, e-3-4-isopropylphenyl acrylic acid, e-3-4-isopropyl-phenyl-acrylic acid, 2e-3-4-isopropylphenyl prop-2-enoic acid
(E)-3-(4-propan-2-ylphenyl)prop-2-enoic acid

Specifications

3368-21-6
C12H14O2
5 g
4-isopropylcinnamic acid, 3-4-isopropylphenyl acrylic acid, p-isopropylcinnamic acid, 2e-3-4-isopropylphenyl acrylic acid, cinnamic acid, p-isopropyl, 2-propenoic acid, 3-4-1-methylethyl phenyl, 2e-3-4-propan-2-yl phenyl prop-2-enoic acid, e-3-4-isopropylphenyl acrylic acid, e-3-4-isopropyl-phenyl-acrylic acid, 2e-3-4-isopropylphenyl prop-2-enoic acid
CC(C)C1=CC=C(\C=C\C(O)=O)C=C1
190.24
190.24
4-Isopropylcinnamic acid, predominantly trans
157°C to 161°C
MFCD00016544
2046679
SJDOOXOUSJDYFE-VMPITWQZSA-N
(E)-3-(4-propan-2-ylphenyl)prop-2-enoic acid
719753
≥98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 222-138-2

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only