CAS RN 3368-21-6
IUPAC Name:
(2E)-3-[4-(propan-2-yl)phenyl]prop-2-enoic acid
Synonyms:
(2E)-3-(4-isopropylphenyl)prop-2-enoic acid
Molecular Weight (g/mol):
190.24
Molecular Formula:
C12H14O2
InChi Key:
SJDOOXOUSJDYFE-VMPITWQZSA-N
SMILES:
CC(C)C1=CC=C(\C=C\C(O)=O)C=C1
1
–
1
of
1
results
1
Promotions Available
4-Isopropylcinnamic acid, predominantly trans, 98+%
4-Isopropylcinnamic acid, predominantly trans, 98+%
CAS: 3368-21-6 Molecular Formula: C12H14O2 Molecular Weight (g/mol): 190.24 MDL Number: MFCD00016544 InChI Key: SJDOOXOUSJDYFE-VMPITWQZSA-N Synonym: 4-isopropylcinnamic acid,3-4-isopropylphenyl acrylic acid,p-isopropylcinnamic acid,2e-3-4-isopropylphenyl acrylic acid,cinnamic acid, p-isopropyl,2-propenoic acid, 3-4-1-methylethyl phenyl,2e-3-4-propan-2-yl phenyl prop-2-enoic acid,e-3-4-isopropylphenyl acrylic acid,e-3-4-isopropyl-phenyl-acrylic acid,2e-3-4-isopropylphenyl prop-2-enoic acid PubChem CID: 719753 IUPAC Name: (E)-3-(4-propan-2-ylphenyl)prop-2-enoic acid SMILES: CC(C)C1=CC=C(\C=C\C(O)=O)C=C1