missing translation for 'onlineSavingsMsg'
Learn More

4-Isopropylcinnamic acid, predominantly trans, 98+%

Catalog No. AAA1515606
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1515606 5 g
AAA1515614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1515606 Supplier Thermo Scientific Chemicals Supplier No. A1515606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3368-21-6 | C12H14O2 | 190.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3368-21-6
Molecular Formula C12H14O2
Molecular Weight (g/mol) 190.24
MDL Number MFCD00016544
InChI Key SJDOOXOUSJDYFE-VMPITWQZSA-N
Synonym 4-isopropylcinnamic acid, 3-4-isopropylphenyl acrylic acid, p-isopropylcinnamic acid, 2e-3-4-isopropylphenyl acrylic acid, cinnamic acid, p-isopropyl, 2-propenoic acid, 3-4-1-methylethyl phenyl, 2e-3-4-propan-2-yl phenyl prop-2-enoic acid, e-3-4-isopropylphenyl acrylic acid, e-3-4-isopropyl-phenyl-acrylic acid, 2e-3-4-isopropylphenyl prop-2-enoic acid
PubChem CID 719753
IUPAC Name (E)-3-(4-propan-2-ylphenyl)prop-2-enoic acid
SMILES CC(C)C1=CC=C(\C=C\C(O)=O)C=C1

Specifications

Melting Point 157°C to 161°C
Quantity 5 g
Beilstein 2046679
Formula Weight 190.24
Percent Purity ≥98%
Chemical Name or Material 4-Isopropylcinnamic acid, predominantly trans
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 222-138-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only