missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Isopropylbenzyl Chloride (contains ca. 10% o-form) 85.0+%, TCI America™
$159.49 - $941.72
Chemical Identifiers
| CAS | 2051-18-5 |
|---|---|
| Molecular Formula | C10H13Cl |
| Molecular Weight (g/mol) | 168.664 |
| MDL Number | MFCD00018885 |
| InChI Key | CYAKWEQUWJAHLW-UHFFFAOYSA-N |
| Synonym | 4-isopropylbenzyl chloride, 1-chloromethyl-4-isopropylbenzene, p-cymene, 7-chloro, p-isopropylbenzyl chloride, 7-chloro-p-cymene, 4-isopropylbenzylchloride, benzene, 1-chloromethyl-4-1-methylethyl, 1-chloromethyl-4-propan-2-yl benzene, p-isopropyl benzyl chloride, benzene, 1-chloromethyl-4-methylethyl |
| PubChem CID | 74916 |
| IUPAC Name | 1-(chloromethyl)-4-propan-2-ylbenzene |
| SMILES | CC(C)C1=CC=C(C=C1)CCl |
Chemical Identifiers
| 2051-18-5 | |
| 168.664 | |
| CYAKWEQUWJAHLW-UHFFFAOYSA-N | |
| 74916 | |
| CC(C)C1=CC=C(C=C1)CCl |
| C10H13Cl | |
| MFCD00018885 | |
| 4-isopropylbenzyl chloride, 1-chloromethyl-4-isopropylbenzene, p-cymene, 7-chloro, p-isopropylbenzyl chloride, 7-chloro-p-cymene, 4-isopropylbenzylchloride, benzene, 1-chloromethyl-4-1-methylethyl, 1-chloromethyl-4-propan-2-yl benzene, p-isopropyl benzyl chloride, benzene, 1-chloromethyl-4-methylethyl | |
| 1-(chloromethyl)-4-propan-2-ylbenzene |
Specifications
| 2051-18-5 | |
| C10H13Cl | |
| 25 g | |
| 4-isopropylbenzyl chloride, 1-chloromethyl-4-isopropylbenzene, p-cymene, 7-chloro, p-isopropylbenzyl chloride, 7-chloro-p-cymene, 4-isopropylbenzylchloride, benzene, 1-chloromethyl-4-1-methylethyl, 1-chloromethyl-4-propan-2-yl benzene, p-isopropyl benzyl chloride, benzene, 1-chloromethyl-4-methylethyl | |
| CC(C)C1=CC=C(C=C1)CCl | |
| 168.664 | |
| 168.66 | |
| Liquid |
| Yellow | |
| MFCD00018885 | |
| 3265 | |
| CYAKWEQUWJAHLW-UHFFFAOYSA-N | |
| 1-(chloromethyl)-4-propan-2-ylbenzene | |
| 74916 | |
| ≥85.0% (GC) | |
| 4-Isopropylbenzyl Chloride (contains ca. 10% o-form) |
Safety and Handling
TSCA : Yes