accessibility menu, dialog, popup

UserName

H3CCH3Cl

CAS RN 2051-18-5

IUPAC Name: 1-(chloromethyl)-4-(propan-2-yl)benzene
Synonyms: 1-(chloromethyl)-4-isopropylbenzene
Molecular Weight (g/mol): 168.66
Molecular Formula: C10H13Cl
InChi Key: CYAKWEQUWJAHLW-UHFFFAOYSA-N
SMILES: CC(C)C1=CC=C(CCl)C=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)