missing translation for 'onlineSavingsMsg'
Learn More

4-Isopropylbenzyl Chloride (contains ca. 10% o-form) 85.0+%, TCI America™

Catalog No. I0225500G
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0225500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I0225500G Supplier TCI America Supplier No. I0225500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2051-18-5
Molecular Formula C10H13Cl
Molecular Weight (g/mol) 168.664
MDL Number MFCD00018885
InChI Key CYAKWEQUWJAHLW-UHFFFAOYSA-N
Synonym 4-isopropylbenzyl chloride, 1-chloromethyl-4-isopropylbenzene, p-cymene, 7-chloro, p-isopropylbenzyl chloride, 7-chloro-p-cymene, 4-isopropylbenzylchloride, benzene, 1-chloromethyl-4-1-methylethyl, 1-chloromethyl-4-propan-2-yl benzene, p-isopropyl benzyl chloride, benzene, 1-chloromethyl-4-methylethyl
PubChem CID 74916
IUPAC Name 1-(chloromethyl)-4-propan-2-ylbenzene
SMILES CC(C)C1=CC=C(C=C1)CCl

Specifications

Color Yellow
Quantity 500 g
UN Number 3265
Formula Weight 168.66
Percent Purity ≥85.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Isopropylbenzyl Chloride (contains ca. 10% o-form)