missing translation for 'onlineSavingsMsg'
Learn More

4-Isopropylbenzyl chloride, 95%

Catalog No. p-7045744
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1183903 1 g
AAL1183906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1183903 Supplier Thermo Scientific Chemicals Supplier No. L1183903

CAS: 2051-18-5 | C10H13Cl | 168.664 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2051-18-5
Molecular Formula C10H13Cl
Molecular Weight (g/mol) 168.664
MDL Number MFCD00018885
InChI Key CYAKWEQUWJAHLW-UHFFFAOYSA-N
Synonym 4-isopropylbenzyl chloride, 1-chloromethyl-4-isopropylbenzene, p-cymene, 7-chloro, p-isopropylbenzyl chloride, 7-chloro-p-cymene, 4-isopropylbenzylchloride, benzene, 1-chloromethyl-4-1-methylethyl, 1-chloromethyl-4-propan-2-yl benzene, p-isopropyl benzyl chloride, benzene, 1-chloromethyl-4-methylethyl
PubChem CID 74916
IUPAC Name 1-(chloromethyl)-4-propan-2-ylbenzene
SMILES CC(C)C1=CC=C(C=C1)CCl

Specifications

Density 1.02
Boiling Point 119°C to 120°C (35 mmHg)
Flash Point 104°C (219°F)
Odor Pungent
Refractive Index 1.523
Quantity 1 g
UN Number UN3265
Beilstein 907207
Formula Weight 168.67
Percent Purity 95%
Chemical Name or Material 4-Isopropylbenzyl chloride
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 218-114-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only