missing translation for 'onlineSavingsMsg'
Learn More

4-Isopropylbenzenethiol 94.0+%, TCI America™

Numéro de catalogue. I059625G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
I059625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. I059625G Fournisseur TCI America Code fournisseur I059625G
Il en reste null

Identifiants chimiques

CAS 4946-14-9
Molecular Formula C9H11S
Molecular Weight (g/mol) 151.25
MDL Number MFCD00041505
InChI Key APDUDRFJNCIWAG-UHFFFAOYSA-M
Synonym 4-isopropylthiophenol, 4-isopropylbenzenethiol, 4-isopropyl thiophenol, 4-propan-2-yl benzene-1-thiol, benzenethiol, 4-1-methylethyl, 4-1-methylethyl benzenethiol, p-cumyl mercaptan, pubchem6814, p-isopropyl thiophenol
PubChem CID 521251
IUPAC Name [4-(propan-2-yl)phenyl]sulfanide
SMILES CC(C)C1=CC=C([S-])C=C1

Spécifications

Color Yellow
Boiling Point 105°C
Quantity 25 g
UN Number 1760
Formula Weight 152.26
Percent Purity ≥94.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Isopropylbenzenethiol