missing translation for 'onlineSavingsMsg'
Learn More

4-Isopropylanisole, 95%

Catalog No. AAA1697014
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1697014 25 g
AAA1697022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1697014 Supplier Thermo Scientific Chemicals Supplier No. A1697014
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4132-48-3 | C10H14O | 150.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4132-48-3
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.22
MDL Number MFCD00015033
InChI Key JULZQKLZSNOEEJ-UHFFFAOYSA-N
Synonym 4-isopropylanisole, 1-isopropyl-4-methoxybenzene, 4-methoxycumene, p-isopropylanisole, anisole, p-isopropyl, 2-4-methoxyphenyl propane, benzene, 1-methoxy-4-1-methylethyl, 1-methoxy-4-propan-2-yl benzene, p-methoxycumene, p-methoxyisopropylbenzene
PubChem CID 77783
IUPAC Name 1-methoxy-4-propan-2-ylbenzene
SMILES COC1=CC=C(C=C1)C(C)C

Specifications

Density 0.935
Boiling Point 212°C to 213°C
Flash Point 83°C (181°F)
Refractive Index 1.503
Quantity 25 g
Beilstein 2080269
Formula Weight 150.22
Percent Purity 95%
Chemical Name or Material 4-Isopropylanisole
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 223-952-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only