missing translation for 'onlineSavingsMsg'
Learn More

4-Isopropylaniline 98.0+%, TCI America™

Catalog No. p-7131367
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I037125ML 25 mL
I0371100ML 100 mL
I0371500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. I037125ML Supplier TCI America Supplier No. I037125ML
Only null left

Chemical Identifiers

CAS 99-88-7
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
MDL Number MFCD00007900
InChI Key LRTFPLFDLJYEKT-UHFFFAOYSA-N
Synonym 4-isopropylaniline, cumidine, 4-aminocumene, p-isopropylaniline, p-cumidine, 4-propan-2-yl aniline, para-isopropylaniline, 4-amino-1-isopropylbenzene, cumene, p-amino, aniline, p-isopropyl
PubChem CID 7464
ChEBI CHEBI:43405
IUPAC Name 4-propan-2-ylaniline
SMILES CC(C)C1=CC=C(C=C1)N

Specifications

Color Yellow
Boiling Point 224°C
Quantity 25 mL
Formula Weight 135.21
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-Isopropylaniline
EINECSNumber (3)-0206
TSCA No