Learn More
4-Isopropylaniline, 99%
CAS: 99-88-7 | C9H13N | 135.21 g/mol
Supplier: Thermo Scientific Chemicals A1464622
Description
4-Isopropylaniline was used in the hydroamination of phenylacetylene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Isopropylaniline was used in the hydroamination of phenylacetylene.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong acids. Light sensitive
Specifications
| 4-Isopropylaniline | |
| -63°C | |
| 226°C to 228°C | |
| C9H13N | |
| MFCD00007900 | |
| 1930544 | |
| 14,2620 | |
| Insoluble in water. | |
| CC(C)C1=CC=C(C=C1)N | |
| 135.21 | |
| CHEBI:43405 | |
| 99% |
| 99-88-7 | |
| 0.989 | |
| 92°C (197°F) | |
| 1.543 | |
| 100 g | |
| Light sensitive | |
| 4-isopropylaniline, cumidine, 4-aminocumene, p-isopropylaniline, p-cumidine, 4-propan-2-yl aniline, para-isopropylaniline, 4-amino-1-isopropylbenzene, cumene, p-amino, aniline, p-isopropyl | |
| LRTFPLFDLJYEKT-UHFFFAOYSA-N | |
| 4-propan-2-ylaniline | |
| 7464 | |
| 135.21 |
Chemical Identifiers
| 99-88-7 | |
| 135.21 | |
| LRTFPLFDLJYEKT-UHFFFAOYSA-N | |
| 7464 | |
| 4-propan-2-ylaniline |
| C9H13N | |
| MFCD00007900 | |
| 4-isopropylaniline, cumidine, 4-aminocumene, p-isopropylaniline, p-cumidine, 4-propan-2-yl aniline, para-isopropylaniline, 4-amino-1-isopropylbenzene, cumene, p-amino, aniline, p-isopropyl | |
| CHEBI:43405 | |
| CC(C)C1=CC=C(C=C1)N |
Safety and Handling
GHS H Statement
H301-H311-H373-H227-H332-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
H227-H302+H312+H332-H315-H319
EINECSNumber : 202-797-2
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only