missing translation for 'onlineSavingsMsg'
Learn More

4-Isopropylaniline, 99%

Catalog No. AAA1464614
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1464614 25 g
AAA1464622 100 g
AAA1464636 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1464614 Supplier Thermo Scientific Chemicals Supplier No. A1464614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 99-88-7 | C9H13N | 135.21 g/mol

4-Isopropylaniline was used in the hydroamination of phenylacetylene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Isopropylaniline was used in the hydroamination of phenylacetylene.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong acids. Light sensitive

Chemical Identifiers

CAS 99-88-7
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
MDL Number MFCD00007900
InChI Key LRTFPLFDLJYEKT-UHFFFAOYSA-N
Synonym 4-isopropylaniline, cumidine, 4-aminocumene, p-isopropylaniline, p-cumidine, 4-propan-2-yl aniline, para-isopropylaniline, 4-amino-1-isopropylbenzene, cumene, p-amino, aniline, p-isopropyl
PubChem CID 7464
ChEBI CHEBI:43405
IUPAC Name 4-propan-2-ylaniline
SMILES CC(C)C1=CC=C(C=C1)N

Specifications

Melting Point -63°C
Density 0.989
Boiling Point 226°C to 228°C
Flash Point 92°C (197°F)
Refractive Index 1.543
Quantity 25 g
Beilstein 1930544
Sensitivity Light sensitive
Merck Index 14,2620
Solubility Information Insoluble in water.
Formula Weight 135.21
Percent Purity 99%
Chemical Name or Material 4-Isopropylaniline
Show More Show Less
Hazard Category H227-H302+H312+H332-H315-H319
Hazard Statement GHS H Statement
H301-H311-H373-H227-H332-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
EINECSNumber 202-797-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only