missing translation for 'onlineSavingsMsg'
Learn More

4-Isopropylaniline, 99%

Catalog No. p-151504
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC174530100 10 g Glass bottle
AC174530500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC174530100 Supplier Thermo Scientific Chemicals Supplier No. 174530100

CAS: 99-88-7 | C9H13N | 135.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 99-88-7
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
MDL Number MFCD00007900
InChI Key LRTFPLFDLJYEKT-UHFFFAOYSA-N
Synonym 4-isopropylaniline, cumidine, 4-aminocumene, p-isopropylaniline, p-cumidine, 4-propan-2-yl aniline, para-isopropylaniline, 4-amino-1-isopropylbenzene, cumene, p-amino, aniline, p-isopropyl
PubChem CID 7464
ChEBI CHEBI:43405
IUPAC Name 4-propan-2-ylaniline
SMILES CC(C)C1=CC=C(C=C1)N

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -63°C
Color Brown to Brown-Red
Density 0.9890g/mL
Boiling Point 226°C to 228°C
Flash Point 92°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5420 to 1.544
Linear Formula (CH3)2CHC6H4NH2
Quantity 10 g
Beilstein 12, 1147
Specific Gravity 0.989
Merck Index 15, 2610
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohols
Formula Weight 135.21
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 4-Isopropylaniline, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 202-797-2
Recommended Storage this product may darken on storage

RUO – Research Use Only