missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Isopropylaminodiphenylamine 98.0+%, TCI America™
$39.14 - $171.49
Chemical Identifiers
| CAS | 101-72-4 |
|---|---|
| Molecular Formula | C15H18N2 |
| Molecular Weight (g/mol) | 226.323 |
| MDL Number | MFCD00026348 |
| InChI Key | OUBMGJOQLXMSNT-UHFFFAOYSA-N |
| Synonym | ippd, n-isopropyl-n'-phenyl-p-phenylenediamine, n-isopropyl-n'-phenyl-1,4-phenylenediamine, cyzone, 4-anilino-n-isopropylaniline, antigen 3c, antioxidant ip, santoflex ip, diaphen fp, cyzone ip |
| PubChem CID | 7573 |
| ChEBI | CHEBI:63569 |
| IUPAC Name | 1-N-phenyl-4-N-propan-2-ylbenzene-1,4-diamine |
| SMILES | CC(C)NC1=CC=C(C=C1)NC2=CC=CC=C2 |
Chemical Identifiers
| 101-72-4 | |
| 226.323 | |
| OUBMGJOQLXMSNT-UHFFFAOYSA-N | |
| 7573 | |
| 1-N-phenyl-4-N-propan-2-ylbenzene-1,4-diamine |
| C15H18N2 | |
| MFCD00026348 | |
| ippd, n-isopropyl-n'-phenyl-p-phenylenediamine, n-isopropyl-n'-phenyl-1,4-phenylenediamine, cyzone, 4-anilino-n-isopropylaniline, antigen 3c, antioxidant ip, santoflex ip, diaphen fp, cyzone ip | |
| CHEBI:63569 | |
| CC(C)NC1=CC=C(C=C1)NC2=CC=CC=C2 |
Specifications
| 101-72-4 | |
| 180°C | |
| MFCD00026348 | |
| 3077 | |
| OUBMGJOQLXMSNT-UHFFFAOYSA-N | |
| 1-N-phenyl-4-N-propan-2-ylbenzene-1,4-diamine | |
| 7573 | |
| 226.32 | |
| Crystalline Powder |
| 80°C | |
| C15H18N2 | |
| 25 g | |
| ippd, n-isopropyl-n'-phenyl-p-phenylenediamine, n-isopropyl-n'-phenyl-1,4-phenylenediamine, cyzone, 4-anilino-n-isopropylaniline, antigen 3c, antioxidant ip, santoflex ip, diaphen fp, cyzone ip | |
| CC(C)NC1=CC=C(C=C1)NC2=CC=CC=C2 | |
| 226.323 | |
| CHEBI:63569 | |
| ≥98.0% (HPLC,T) | |
| 4-Isopropylaminodiphenylamine |
Safety and Handling
EINECSNumber : (3)-0136&(3)-0368
RTECSNumber : ST2650000
TSCA : Yes