accessibility menu, dialog, popup

UserName

CH3H3CNHNH

CAS RN 101-72-4

IUPAC Name: N1-phenyl-N4-(propan-2-yl)benzene-1,4-diamine
Synonyms: IPPD -isopropylaminodiphenylamine
Molecular Weight (g/mol): 226.32
Molecular Formula: C15H18N2
InChi Key: OUBMGJOQLXMSNT-UHFFFAOYSA-N
SMILES: CC(C)NC1=CC=C(NC2=CC=CC=C2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results