missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4'-Isopropylacetophenone 95.0+%, TCI America™
Supplier: TCI America I057025ML
Specifications
| 4′-Isopropylacetophenone | |
| Yellow | |
| C11H14O | |
| 25 mL | |
| PDLCCNYKIIUWHA-UHFFFAOYSA-N | |
| 1-[4-(propan-2-yl)phenyl]ethan-1-one | |
| 12578 | |
| ≥95.0% (GC) |
| 645-13-6 | |
| 238°C | |
| MFCD00048297 | |
| 4'-isopropylacetophenone, 4-isopropylacetophenone, p-isopropylacetophenone, cuminone, 1-4-isopropylphenyl ethanone, ethanone, 1-4-1-methylethyl phenyl, acetophenone, 4'-isopropyl, unii-742zkz2a6d, methyl p-isopropylphenyl ketone, 1-4-propan-2-yl phenyl ethan-1-one | |
| CC(C)C1=CC=C(C=C1)C(C)=O | |
| 162.23 | |
| 162.23 | |
| Liquid |
Chemical Identifiers
| 645-13-6 | |
| 162.23 | |
| PDLCCNYKIIUWHA-UHFFFAOYSA-N | |
| 12578 | |
| CC(C)C1=CC=C(C=C1)C(C)=O |
| C11H14O | |
| MFCD00048297 | |
| 4'-isopropylacetophenone, 4-isopropylacetophenone, p-isopropylacetophenone, cuminone, 1-4-isopropylphenyl ethanone, ethanone, 1-4-1-methylethyl phenyl, acetophenone, 4'-isopropyl, unii-742zkz2a6d, methyl p-isopropylphenyl ketone, 1-4-propan-2-yl phenyl ethan-1-one | |
| 1-[4-(propan-2-yl)phenyl]ethan-1-one |
Safety and Handling
TSCA : Yes