missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Isopropyl-4'-methyldiphenyliodonium Tetrakis(pentafluorophenyl)borate 98.0+%, TCI America™
$365.35 - $1,247.05
Chemical Identifiers
| CAS | 178233-72-2 |
|---|---|
| Molecular Formula | C40H18BF20I |
| Molecular Weight (g/mol) | 1016.267 |
| MDL Number | MFCD03411231 |
| InChI Key | MJDBKUFRQPPWGN-UHFFFAOYSA-N |
| Synonym | p-Cumenyl(p-tolyl)iodonium Tetrakis(pentafluorophenyl)borate |
| PubChem CID | 21967041 |
| IUPAC Name | (4-methylphenyl)-(4-propan-2-ylphenyl)iodanium;tetrakis(2,3,4,5,6-pentafluorophenyl)boranuide |
| SMILES | [B-](C1=C(C(=C(C(=C1F)F)F)F)F)(C2=C(C(=C(C(=C2F)F)F)F)F)(C3=C(C(=C(C(=C3F)F)F)F)F)C4=C(C(=C(C(=C4F)F)F)F)F.CC1=CC=C(C=C1)[I+]C2=CC=C(C=C2)C(C)C |
Chemical Identifiers
| 178233-72-2 | |
| 1016.267 | |
| MJDBKUFRQPPWGN-UHFFFAOYSA-N | |
| 21967041 | |
| [B-](C1=C(C(=C(C(=C1F)F)F)F)F)(C2=C(C(=C(C(=C2F)F)F)F)F)(C3=C(C(=C(C(=C3F)F)F)F)F)C4=C(C(=C(C(=C4F)F)F)F)F.CC1=CC=C(C=C1)[I+]C2=CC=C(C=C2)C(C)C |
| C40H18BF20I | |
| MFCD03411231 | |
| p-Cumenyl(p-tolyl)iodonium Tetrakis(pentafluorophenyl)borate | |
| (4-methylphenyl)-(4-propan-2-ylphenyl)iodanium;tetrakis(2,3,4,5,6-pentafluorophenyl)boranuide |
Specifications
| 178233-72-2 | |
| White-Yellow | |
| MFCD03411231 | |
| 3077 | |
| MJDBKUFRQPPWGN-UHFFFAOYSA-N | |
| (4-methylphenyl)-(4-propan-2-ylphenyl)iodanium;tetrakis(2,3,4,5,6-pentafluorophenyl)boranuide | |
| 21967041 | |
| ≥98.0% (T) | |
| 4-Isopropyl-4′-methyldiphenyliodonium Tetrakis(pentafluorophenyl)borate |
| 133°C | |
| C40H18BF20I | |
| 5 g | |
| p-Cumenyl(p-tolyl)iodonium Tetrakis(pentafluorophenyl)borate | |
| [B-](C1=C(C(=C(C(=C1F)F)F)F)F)(C2=C(C(=C(C(=C2F)F)F)F)F)(C3=C(C(=C(C(=C3F)F)F)F)F)C4=C(C(=C(C(=C4F)F)F)F)F.CC1=CC=C(C=C1)[I+]C2=CC=C(C=C2)C(C)C | |
| 1016.267 | |
| 1016.27 | |
| Crystalline Powder |
Safety and Handling
TSCA : Yes