accessibility menu, dialog, popup

UserName

CAS RN 178233-72-2

BFFFFFFFFFFFFFFFFFFFFCH3ICH3H3C

CAS RN 178233-72-2

IUPAC Name: (4-methylphenyl)[4-(propan-2-yl)phenyl]iodanium; tetrakis(2,3,4,5,6-pentafluorophenyl)boranuide
Synonyms: (4-isopropylphenyl)(4-methylphenyl)iodanium; tetrakis(2,3,4,5,6-pentafluorophenyl)boranuide
Molecular Weight (g/mol): 1016.27
Molecular Formula: C40H18BF20I
InChi Key: MJDBKUFRQPPWGN-UHFFFAOYSA-N
SMILES: CC(C)C1=CC=C([I+]C2=CC=C(C)C=C2)C=C1.FC1=C(F)C(F)=C(C(F)=C1F)[B-](C1=C(F)C(F)=C(F)C(F)=C1F)(C1=C(F)C(F)=C(F)C(F)=C1F)C1=C(F)C(F)=C(F)C(F)=C1F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results