missing translation for 'onlineSavingsMsg'
Learn More

4-Isopropyl-4'-methyldiphenyliodonium Tetrakis(pentafluorophenyl)borate 98.0+%, TCI America™

Catalog No. I05915G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I05915G 5 g
I059125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I05915G Supplier TCI America Supplier No. I05915G
Only null left

Chemical Identifiers

CAS 178233-72-2
Molecular Formula C40H18BF20I
Molecular Weight (g/mol) 1016.267
MDL Number MFCD03411231
InChI Key MJDBKUFRQPPWGN-UHFFFAOYSA-N
Synonym p-Cumenyl(p-tolyl)iodonium Tetrakis(pentafluorophenyl)borate
PubChem CID 21967041
IUPAC Name (4-methylphenyl)-(4-propan-2-ylphenyl)iodanium;tetrakis(2,3,4,5,6-pentafluorophenyl)boranuide
SMILES [B-](C1=C(C(=C(C(=C1F)F)F)F)F)(C2=C(C(=C(C(=C2F)F)F)F)F)(C3=C(C(=C(C(=C3F)F)F)F)F)C4=C(C(=C(C(=C4F)F)F)F)F.CC1=CC=C(C=C1)[I+]C2=CC=C(C=C2)C(C)C

Specifications

Melting Point 133°C
Color White-Yellow
Quantity 5 g
UN Number 3077
Formula Weight 1016.27
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Isopropyl-4′-methyldiphenyliodonium Tetrakis(pentafluorophenyl)borate