missing translation for 'onlineSavingsMsg'
Learn More

4-Isopropyl-3-methylphenol 99.0+%, TCI America™

Catalog No. p-7132149
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M040925G 25 g
M0409100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M040925G Supplier TCI America Supplier No. M040925G
Only null left

Chemical Identifiers

CAS 3228-02-2
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.22
MDL Number MFCD00010704
InChI Key IJALWSVNUBBQRA-UHFFFAOYSA-N
Synonym Biosol, 4-Isopropyl-m-cresol
PubChem CID 18597
ChEBI CHEBI:34429
IUPAC Name 3-methyl-4-propan-2-ylphenol
SMILES CC1=C(C=CC(=C1)O)C(C)C

Specifications

Melting Point 112°C
Color White-Yellow
Boiling Point 246°C
Quantity 25 g
Formula Weight 150.22
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Isopropyl-3-methylphenol
EINECSNumber (3)-0521&(4)-0057
RTECSNumber GZ7170000
TSCA No