missing translation for 'onlineSavingsMsg'
Learn More

4-Isopropoxybenzenesulfonyl Chloride 98.0+%, TCI America™

Catalog No. p-7131603
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I08025G 5 g
I080225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I08025G Supplier TCI America Supplier No. I08025G
Only null left

Chemical Identifiers

CAS 98995-40-5
Molecular Formula C9H11ClO3S
Molecular Weight (g/mol) 234.694
MDL Number MFCD03093083
InChI Key IJWCRHKAQNFJLT-UHFFFAOYSA-N
Synonym 4-isopropoxybenzenesulfonyl chloride, 4-isopropoxybenzene-1-sulfonyl chloride, 4-isopropoxyphenylsulfonyl chloride, 4-propan-2-yloxy benzene-1-sulfonyl chloride, benzenesulfonyl chloride, 4-1-methylethoxy, 4-isopropoxybenzenesulphonyl chloride, benzenesulfonyl chloride, 4-1-methylethoxy-9ci, 4-isopropoxybenzenesulfonylchloride, pubchem19078, acmc-209sbk
PubChem CID 22327297
IUPAC Name 4-propan-2-yloxybenzenesulfonyl chloride
SMILES CC(C)OC1=CC=C(C=C1)S(=O)(=O)Cl

Specifications

Melting Point 38°C
Boiling Point 113°C
Quantity 5 g
UN Number 3261
Formula Weight 234.69
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Isopropoxybenzenesulfonyl Chloride
TSCA No
Recommended Storage Refrigerator