accessibility menu, dialog, popup

UserName

CH3H3COSOOCl

CAS RN 98995-40-5

IUPAC Name: 4-(propan-2-yloxy)benzene-1-sulfonyl chloride
Synonyms: 4-isopropoxybenzenesulfonyl chloride
Molecular Weight (g/mol): 234.69
Molecular Formula: C9H11ClO3S
InChi Key: IJWCRHKAQNFJLT-UHFFFAOYSA-N
SMILES: CC(C)OC1=CC=C(C=C1)S(Cl)(=O)=O
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)