missing translation for 'onlineSavingsMsg'
Learn More

4-Isopropoxybenzeneboronic acid, 97%

Catalog No. p-7034973
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5267503 1 g
AAH5267506 5 g
AAH5267514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH5267503 Supplier Thermo Scientific Chemicals Supplier No. H5267503
Only null left

CAS: 153624-46-5 | C9H13BO3 | 180.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 153624-46-5
Molecular Formula C9H13BO3
Molecular Weight (g/mol) 180.01
MDL Number MFCD03427051
InChI Key CJUHQADBFQRIMC-UHFFFAOYSA-N
Synonym 4-isopropoxyphenylboronic acid, 4-isopropoxylphenylboronic acid, 4-isopropoxyphenyl boronic acid, 4-isopropoxybenzeneboronic acid, 4-isopropyloxyphenylboronic acid, 4-propan-2-yloxyphenyl boronic acid, boronic acid, 4-1-methylethoxy phenyl, 4-propan-2-yloxy phenyl boronic acid, 4-iso-propoxyphenylboronic acid, p-isopropoxyphenylboronic acid
PubChem CID 3698726
IUPAC Name (4-propan-2-yloxyphenyl)boronic acid
SMILES B(C1=CC=C(C=C1)OC(C)C)(O)O

Specifications

Melting Point 150°C to 154°C
Quantity 1 g
Formula Weight 180.01
Percent Purity 97%
Chemical Name or Material 4-Isopropoxybenzeneboronic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only