missing translation for 'onlineSavingsMsg'
Learn More

4-Isopropoxyaniline 98.0+%, TCI America™

Catalog No. p-7131386
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I03995G 5 g
I039925G 25 g
I0399100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. I03995G Supplier TCI America Supplier No. I03995G
Only null left

Chemical Identifiers

CAS 7664-66-6
Molecular Formula C9H13NO
Molecular Weight (g/mol) 151.209
MDL Number MFCD00059188
InChI Key MLNFMFAMNBGAQT-UHFFFAOYSA-N
PubChem CID 82112
IUPAC Name 4-propan-2-yloxyaniline
SMILES CC(C)OC1=CC=C(C=C1)N

Specifications

Color Yellow
Boiling Point 95°C
Quantity 5 g
Formula Weight 151.21
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-Isopropoxyaniline