missing translation for 'onlineSavingsMsg'
Learn More

4-Isopropoxy-2-butanone 98.0+%, TCI America™

Catalog No. I02835ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I02835ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I02835ML Supplier TCI America Supplier No. I02835ML
Only null left

Chemical Identifiers

CAS 32541-58-5
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.19
MDL Number MFCD00059358
InChI Key MEWOUXCWMNHTIZ-UHFFFAOYSA-N
PubChem CID 538375
IUPAC Name 4-(propan-2-yloxy)butan-2-one
SMILES CC(C)OCCC(C)=O

Specifications

Color Yellow
Boiling Point 75°C
Quantity 5 mL
UN Number 1993
Formula Weight 130.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Isopropoxy-2-butanone