accessibility menu, dialog, popup

UserName

H3CCH3OOCH3

CAS RN 32541-58-5

IUPAC Name: 4-(propan-2-yloxy)butan-2-one
Synonyms: 4-isopropoxybutan-2-one
Molecular Weight (g/mol): 130.19
Molecular Formula: C7H14O2
InChi Key: MEWOUXCWMNHTIZ-UHFFFAOYSA-N
SMILES: CC(C)OCCC(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results