Learn More
4-Isocyanatobenzeneboronic acid pinacol ester, 95%
CAS: 380430-64-8 | C13H16BNO3 | 245.085 g/mol
Supplier: Thermo Scientific Chemicals H5300303
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 4-Isocyanatobenzeneboronic acid pinacol ester | |
| 162°C to 168°C | |
| MFCD03411942 | |
| 2-4-isocyanatophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-isocyanatophenylboronic acid pinacol ester, 1,3,2-dioxaborolane, 2-4-isocyanatophenyl-4,4,5,5-tetramethyl, 4-isocyanatophenylboronic acid, pinacol ester, 4-isocyanatophenyl boronic acid pinacol ester, 4-isocyanatobenzeneboronic acid pinacol ester, 4-isocyanatophenyl boronic acid, pinacol ester, 4-isocyanatophenyl boronic acid,pinacol ester, 2-4-isocyanato-phenyl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane | |
| B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)N=C=O | |
| 245.085 | |
| 245.08 |
| 380430-64-8 | |
| C13H16BNO3 | |
| 1 g | |
| RHKRIDVZGAXYQE-UHFFFAOYSA-N | |
| 2-(4-isocyanatophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane | |
| 2773474 | |
| 95% |
Chemical Identifiers
| 380430-64-8 | |
| 245.085 | |
| RHKRIDVZGAXYQE-UHFFFAOYSA-N | |
| 2773474 | |
| B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)N=C=O |
| C13H16BNO3 | |
| MFCD03411942 | |
| 2-4-isocyanatophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-isocyanatophenylboronic acid pinacol ester, 1,3,2-dioxaborolane, 2-4-isocyanatophenyl-4,4,5,5-tetramethyl, 4-isocyanatophenylboronic acid, pinacol ester, 4-isocyanatophenyl boronic acid pinacol ester, 4-isocyanatobenzeneboronic acid pinacol ester, 4-isocyanatophenyl boronic acid, pinacol ester, 4-isocyanatophenyl boronic acid,pinacol ester, 2-4-isocyanato-phenyl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane | |
| 2-(4-isocyanatophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane |
Safety and Handling
GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only