missing translation for 'onlineSavingsMsg'
Learn More

4-Isocyanatobenzeneboronic acid pinacol ester, 95%

Catalog No. p-7035289
Change view
Click to view available options
Quantity:
250 mg
1 g
Catalog No. Quantity
AAH53003MD 250 mg
AAH5300303 1 g
2 options

Catalog No. AAH53003MD

Supplier: Thermo Scientific Chemicals H53003MD

Only null left

CAS: 380430-64-8 | C13H16BNO3 | 245.085 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 380430-64-8
Molecular Formula C13H16BNO3
Molecular Weight (g/mol) 245.085
MDL Number MFCD03411942
InChI Key RHKRIDVZGAXYQE-UHFFFAOYSA-N
Synonym 2-4-isocyanatophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-isocyanatophenylboronic acid pinacol ester, 1,3,2-dioxaborolane, 2-4-isocyanatophenyl-4,4,5,5-tetramethyl, 4-isocyanatophenylboronic acid, pinacol ester, 4-isocyanatophenyl boronic acid pinacol ester, 4-isocyanatobenzeneboronic acid pinacol ester, 4-isocyanatophenyl boronic acid, pinacol ester, 4-isocyanatophenyl boronic acid,pinacol ester, 2-4-isocyanato-phenyl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane
PubChem CID 2773474
IUPAC Name 2-(4-isocyanatophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)N=C=O

Specifications

Melting Point 162°C to 168°C
Quantity 250 mg
Formula Weight 245.08
Percent Purity 95%
Chemical Name or Material 4-Isocyanatobenzeneboronic acid pinacol ester
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only