missing translation for 'onlineSavingsMsg'
Learn More

4-Isobutyrylacetophenone Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111002435
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002435 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002435 Supplier Merck Emd Millipore Supplier No. PHR2124100MG

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Specifications

CAS 103931-20-0
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C12H14O2
Type Certified Reference Material
Quantity 100 mg
Formula Weight 190.24
Grade Certified Reference Material