missing translation for 'onlineSavingsMsg'
Learn More

4-Isobutylcyclohexanecarboxylic Acid (cis- and trans- mixture) 98.0+%, TCI America™

Supplier:  TCI America I08735G

Catalog No. I08735G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

4-Isobutylcyclohexanecarboxylic Acid (cis- and trans- mixture)
White
5 g
CC(C)CC1CCC(CC1)C(=O)O
184.279
184.28
Crystalline Lumps
38792-88-0
C11H20O2
NFJJRCCYQXONBL-UHFFFAOYSA-N
4-(2-methylpropyl)cyclohexane-1-carboxylic acid
217396
≥98.0% (GC,T)

Chemical Identifiers

38792-88-0
184.279
217396
CC(C)CC1CCC(CC1)C(=O)O
C11H20O2
NFJJRCCYQXONBL-UHFFFAOYSA-N
4-(2-methylpropyl)cyclohexane-1-carboxylic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : GU8584000

TSCA : No

SDS
Documents

Documents

Promotions

Promotions