accessibility menu, dialog, popup

UserName

CAS RN 38792-88-0

CH3H3COHO

CAS RN 38792-88-0

IUPAC Name: 4-(2-methylpropyl)cyclohexane-1-carboxylic acid
Synonyms: 4-(2-methylpropyl)cyclohexane-1-carboxylic acid
Molecular Weight (g/mol): 184.28
Molecular Formula: C11H20O2
InChi Key: NFJJRCCYQXONBL-UHFFFAOYSA-N
SMILES: CC(C)CC1CCC(CC1)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results