Learn More
4-Isobutylbenzeneboronic acid, 98%
CAS: 153624-38-5 | C10H15BO2 | 178.038 g/mol
$56.33 - $217.14
Chemical Identifiers
| CAS | 153624-38-5 |
|---|---|
| Molecular Formula | C10H15BO2 |
| Molecular Weight (g/mol) | 178.038 |
| MDL Number | MFCD01632198 |
| InChI Key | YZSPHVWALUJQNK-UHFFFAOYSA-N |
| Synonym | 4-isobutylphenylboronic acid, 4-isobutylbenzeneboronic acid, 4-isobutylphenyl boronic acid, 4-2-methylpropyl phenylboronic acid, 4-2-methylpropyl phenyl boronic acid, 4-2-methylpropyl phenyl boranediol, boronic acid, 4-2-methylpropyl phenyl, boronic acid,b-4-2-methylpropyl phenyl, 4-iso-butylphenylboronic acid, pubchem20832 |
| PubChem CID | 2773472 |
| IUPAC Name | [4-(2-methylpropyl)phenyl]boronic acid |
| SMILES | B(C1=CC=C(C=C1)CC(C)C)(O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH5260303
|
Thermo Scientific Chemicals
H5260303 |
1 g |
Each for $56.33
|
|
|||||
|
AAH5260306
|
Thermo Scientific Chemicals
H5260306 |
5 g |
Each for $217.14
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 153624-38-5 | |
| 178.038 | |
| YZSPHVWALUJQNK-UHFFFAOYSA-N | |
| 2773472 | |
| B(C1=CC=C(C=C1)CC(C)C)(O)O |
| C10H15BO2 | |
| MFCD01632198 | |
| 4-isobutylphenylboronic acid, 4-isobutylbenzeneboronic acid, 4-isobutylphenyl boronic acid, 4-2-methylpropyl phenylboronic acid, 4-2-methylpropyl phenyl boronic acid, 4-2-methylpropyl phenyl boranediol, boronic acid, 4-2-methylpropyl phenyl, boronic acid,b-4-2-methylpropyl phenyl, 4-iso-butylphenylboronic acid, pubchem20832 | |
| [4-(2-methylpropyl)phenyl]boronic acid |
Specifications
| 153624-38-5 | |
| C10H15BO2 | |
| 1 g | |
| YZSPHVWALUJQNK-UHFFFAOYSA-N | |
| [4-(2-methylpropyl)phenyl]boronic acid | |
| 2773472 | |
| 98% |
| 117°C to 120°C | |
| MFCD01632198 | |
| 4-isobutylphenylboronic acid, 4-isobutylbenzeneboronic acid, 4-isobutylphenyl boronic acid, 4-2-methylpropyl phenylboronic acid, 4-2-methylpropyl phenyl boronic acid, 4-2-methylpropyl phenyl boranediol, boronic acid, 4-2-methylpropyl phenyl, boronic acid,b-4-2-methylpropyl phenyl, 4-iso-butylphenylboronic acid, pubchem20832 | |
| B(C1=CC=C(C=C1)CC(C)C)(O)O | |
| 178.038 | |
| 178.04 | |
| 4-Isobutylbenzeneboronic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only