missing translation for 'onlineSavingsMsg'
Learn More

4-Isobutylbenzeneboronic acid, 98%

Catalog No. AAH5260306
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5260306 5 g
AAH5260303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5260306 Supplier Thermo Scientific Chemicals Supplier No. H5260306

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 153624-38-5 | C10H15BO2 | 178.038 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 153624-38-5
Molecular Formula C10H15BO2
Molecular Weight (g/mol) 178.038
MDL Number MFCD01632198
InChI Key YZSPHVWALUJQNK-UHFFFAOYSA-N
Synonym 4-isobutylphenylboronic acid, 4-isobutylbenzeneboronic acid, 4-isobutylphenyl boronic acid, 4-2-methylpropyl phenylboronic acid, 4-2-methylpropyl phenyl boronic acid, 4-2-methylpropyl phenyl boranediol, boronic acid, 4-2-methylpropyl phenyl, boronic acid,b-4-2-methylpropyl phenyl, 4-iso-butylphenylboronic acid, pubchem20832
PubChem CID 2773472
IUPAC Name [4-(2-methylpropyl)phenyl]boronic acid
SMILES B(C1=CC=C(C=C1)CC(C)C)(O)O

Specifications

Melting Point 117°C to 120°C
Quantity 5 g
Formula Weight 178.04
Percent Purity 98%
Chemical Name or Material 4-Isobutylbenzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only