missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4'-Isobutylacetophenone 96.0+%, TCI America™
Supplier: TCI America I041725ML
Specifications
| 4′-Isobutylacetophenone | |
| Yellow | |
| C12H16O | |
| 25 mL | |
| KEAGRYYGYWZVPC-UHFFFAOYSA-N | |
| 1-[4-(2-methylpropyl)phenyl]ethanone | |
| 93214 | |
| ≥96.0% (GC) |
| 38861-78-8 | |
| 268°C | |
| MFCD00027393 | |
| 4'-isobutylacetophenone, 4-isobutylacetophenone, p-iso-butylacetophenone, 1-4-isobutylphenyl ethanone, p-isobutylacetophenone, ethanone, 1-4-2-methylpropyl phenyl, 1-4-2-methylpropyl phenyl ethan-1-one, acetophenone, 4-isobutyl, 1-4-2-methylpropyl phenyl ethanone, unii-aml715rd20 | |
| CC(C)CC1=CC=C(C=C1)C(=O)C | |
| 176.259 | |
| 176.26 | |
| Liquid |
Chemical Identifiers
| 38861-78-8 | |
| 176.259 | |
| KEAGRYYGYWZVPC-UHFFFAOYSA-N | |
| 93214 | |
| CC(C)CC1=CC=C(C=C1)C(=O)C |
| C12H16O | |
| MFCD00027393 | |
| 4'-isobutylacetophenone, 4-isobutylacetophenone, p-iso-butylacetophenone, 1-4-isobutylphenyl ethanone, p-isobutylacetophenone, ethanone, 1-4-2-methylpropyl phenyl, 1-4-2-methylpropyl phenyl ethan-1-one, acetophenone, 4-isobutyl, 1-4-2-methylpropyl phenyl ethanone, unii-aml715rd20 | |
| 1-[4-(2-methylpropyl)phenyl]ethanone |
Safety and Handling
EINECSNumber : (3)-2928
TSCA : Yes