missing translation for 'onlineSavingsMsg'
Learn More

4'-Isobutylacetophenone 96.0+%, TCI America™

Catalog No. p-7131398
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I041725ML 25 mL
I0417100ML 100 mL
I0417500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. I041725ML Supplier TCI America Supplier No. I041725ML
Only null left

Chemical Identifiers

CAS 38861-78-8
Molecular Formula C12H16O
Molecular Weight (g/mol) 176.259
MDL Number MFCD00027393
InChI Key KEAGRYYGYWZVPC-UHFFFAOYSA-N
Synonym 4'-isobutylacetophenone, 4-isobutylacetophenone, p-iso-butylacetophenone, 1-4-isobutylphenyl ethanone, p-isobutylacetophenone, ethanone, 1-4-2-methylpropyl phenyl, 1-4-2-methylpropyl phenyl ethan-1-one, acetophenone, 4-isobutyl, 1-4-2-methylpropyl phenyl ethanone, unii-aml715rd20
PubChem CID 93214
IUPAC Name 1-[4-(2-methylpropyl)phenyl]ethanone
SMILES CC(C)CC1=CC=C(C=C1)C(=O)C

Specifications

Color Yellow
Boiling Point 268°C
Quantity 25 mL
Formula Weight 176.26
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 4′-Isobutylacetophenone
EINECSNumber (3)-2928
TSCA Yes