missing translation for 'onlineSavingsMsg'
Learn More

4'-Isobutylacetophenone, 97%

CAS: 38861-78-8 | C12H16O | 176.26 g/mol

Supplier:  Thermo Scientific Chemicals 448820500

 View more versions of this product

Catalog No. AC448820500


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4'-Isobutylacetophenone
0.9600g/mL
54°C
Glass Bottle
(CH3)2CHCH2C6H4COCH3
0.96
KEAGRYYGYWZVPC-UHFFFAOYSA-N
1-[4-(2-methylpropyl)phenyl]ethanone
93214
97%
38861-78-8
107.0°C (2.0 mmHg)
96% min. (GC)
C12H16O
50 g
4'-isobutylacetophenone, 4-isobutylacetophenone, p-iso-butylacetophenone, 1-4-isobutylphenyl ethanone, p-isobutylacetophenone, ethanone, 1-4-2-methylpropyl phenyl, 1-4-2-methylpropyl phenyl ethan-1-one, acetophenone, 4-isobutyl, 1-4-2-methylpropyl phenyl ethanone, unii-aml715rd20
CC(C)CC1=CC=C(C=C1)C(=O)C
176.26
176.26

Chemical Identifiers

38861-78-8
176.26
4'-isobutylacetophenone, 4-isobutylacetophenone, p-iso-butylacetophenone, 1-4-isobutylphenyl ethanone, p-isobutylacetophenone, ethanone, 1-4-2-methylpropyl phenyl, 1-4-2-methylpropyl phenyl ethan-1-one, acetophenone, 4-isobutyl, 1-4-2-methylpropyl phenyl ethanone, unii-aml715rd20
1-[4-(2-methylpropyl)phenyl]ethanone
C12H16O
KEAGRYYGYWZVPC-UHFFFAOYSA-N
93214
CC(C)CC1=CC=C(C=C1)C(=O)C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapor.
May cause an allergic skin reaction.
Toxic to aquatic life with long lasting effects.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rin

GHS Signal Word: Warning

EINECSNumber : 254-159-8

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only