missing translation for 'onlineSavingsMsg'
Learn More

4-Iodotoluene, 98%

Catalog No. p-7022731
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1563114 25 g
AAA1563122 100 g
AAA1563136 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1563114 Supplier Thermo Scientific Chemicals Supplier No. A1563114

CAS: 624-31-7 | C7H7I | 218.037 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
4-Iodotoluene is used in wide range of medicals industrial applications as well as in human and animal nutrition products such as antiseptics and disinfectants, pharmaceutical intermediates, polarizing films for liquid crystal display [LCD] chemicals.

Solubility
Solubility in methanol is almost transparent.

Notes
Light sensitive. Store in dark. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 624-31-7
Molecular Formula C7H7I
Molecular Weight (g/mol) 218.037
MDL Number MFCD00001059
InChI Key UDHAWRUAECEBHC-UHFFFAOYSA-N
Synonym 4-iodotoluene, p-iodotoluene, benzene, 1-iodo-4-methyl, toluene, p-iodo, p-tolyl iodide, p-methyliodobenzene, 1-methyl-4-iodobenzene, 1-iodo-4-methyl-benzene, unii-g75kcn1am8, iodotoluene 4-
PubChem CID 12207
IUPAC Name 1-iodo-4-methylbenzene
SMILES CC1=CC=C(C=C1)I

Specifications

Melting Point 33°C to 35°C
Density 1.678
Boiling Point 210°C to 212°C
Flash Point 90°C (194°F)
Quantity 25 g
Beilstein 1903637
Sensitivity Light sensitive
Formula Weight 218.04
Percent Purity 98%
Chemical Name or Material 4-Iodotoluene
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 210-841-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only