missing translation for 'onlineSavingsMsg'
Learn More

4-Iodotoluene, 98%

Catalog No. p-149559
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC133940100 10 g Glass Bottle
AC133940500 50 g Glass Bottle
AC133941000 100 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC133940100 Supplier Thermo Scientific Chemicals Supplier No. 133940100
Only null left

CAS: 624-31-7 | C7H7I | 218.037 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 624-31-7
Molecular Formula C7H7I
Molecular Weight (g/mol) 218.037
MDL Number MFCD00001059
InChI Key UDHAWRUAECEBHC-UHFFFAOYSA-N
Synonym 4-iodotoluene, p-iodotoluene, benzene, 1-iodo-4-methyl, toluene, p-iodo, p-tolyl iodide, p-methyliodobenzene, 1-methyl-4-iodobenzene, 1-iodo-4-methyl-benzene, unii-g75kcn1am8, iodotoluene 4-
PubChem CID 12207
IUPAC Name 1-iodo-4-methylbenzene
SMILES CC1=CC=C(C=C1)I

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 33°C to 36°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Linear Formula IC6H4CH3
Quantity 10 g
Beilstein 05, 312
Formula Weight 218.04
Percent Purity 98%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material 4-Iodotoluene, 98%
Show More Show Less
EINECSNumber 210-841-7
TSCA TSCA

RUO – Research Use Only