missing translation for 'onlineSavingsMsg'
Learn More

4-Iodotetrahydrothiopyran, 97%, Thermo Scientific™

Catalog No. CC61911DA
Change view
Click to view available options
Quantity:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC61911DA 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC61911DA Supplier Thermo Scientific Chemicals Supplier No. CC61911DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 281204-90-8
Molecular Formula C5H9IS
Molecular Weight (g/mol) 228.09
MDL Number MFCD09025908
InChI Key JVBIONNYIWEILC-UHFFFAOYSA-N
Synonym 4-iodotetrahydrothiopyran, 4-iodotetrahydro-2h-thiopyran, 2h-thiopyran,tetrahydro-4-iodo
PubChem CID 18525911
IUPAC Name 4-iodothiane
SMILES IC1CCSCC1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 1g
Formula Weight 228.09
Percent Purity 97%
Chemical Name or Material 4-Iodotetrahydrothiopyran
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.