accessibility menu, dialog, popup

UserName

CAS RN 281204-90-8

IUPAC Name: 4-iodothiane
Synonyms: 4-iodothiane
Molecular Weight (g/mol): 228.09
Molecular Formula: C5H9IS
InChi Key: JVBIONNYIWEILC-UHFFFAOYSA-N
SMILES: IC1CCSCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)