missing translation for 'onlineSavingsMsg'
Learn More

4-Iodophenylacetic acid, 97%

Catalog No. AC450900010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC450900010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC450900010 Supplier Thermo Scientific Chemicals Supplier No. 450900010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1798-06-7 | C8H7IO2 | 262.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1798-06-7
Molecular Formula C8H7IO2
Molecular Weight (g/mol) 262.05
InChI Key FJSHTWVDFAUNCO-UHFFFAOYSA-N
Synonym 4-iodophenylacetic acid, 2-4-iodophenyl acetic acid, 4-iodophenyl acetic acid, 4-iodophenylaceticacid, p-iodophenylacetic acid, benzeneacetic acid, 4-iodo, 4-iodo-phenyl-acetic acid, pubchem18835, 4-iodoohenylacetic acid, 4-iodobenzeneacetic acid
PubChem CID 137214
IUPAC Name 2-(4-iodophenyl)acetic acid
SMILES C1=CC(=CC=C1CC(=O)O)I

Specifications

Melting Point 140°C to 141.5°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 262.05
Percent Purity 97%
Chemical Name or Material 4-Iodophenylacetic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

RUO – Research Use Only