missing translation for 'onlineSavingsMsg'
Learn More

4-Iodophenylacetic acid, 97%

Catalog No. AAL1334506
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1334506 5 g
AAL1334503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1334506 Supplier Thermo Scientific Chemicals Supplier No. L1334506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1798-06-7 | C8H7IO2 | 262.046 g/mol

4-Iodophenylacetic acid, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used in suzuki coupling reaction transport.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Iodophenylacetic acid, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used in suzuki coupling reaction transport.

Solubility
Insoluble in water.

Notes
Light Sensitive. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.

Chemical Identifiers

CAS 1798-06-7
Molecular Formula C8H7IO2
Molecular Weight (g/mol) 262.046
MDL Number MFCD00082985
InChI Key FJSHTWVDFAUNCO-UHFFFAOYSA-N
Synonym 4-iodophenylacetic acid, 2-4-iodophenyl acetic acid, 4-iodophenyl acetic acid, 4-iodophenylaceticacid, p-iodophenylacetic acid, benzeneacetic acid, 4-iodo, 4-iodo-phenyl-acetic acid, pubchem18835, 4-iodoohenylacetic acid, 4-iodobenzeneacetic acid
PubChem CID 137214
IUPAC Name 2-(4-iodophenyl)acetic acid
SMILES C1=CC(=CC=C1CC(=O)O)I

Specifications

Melting Point 136°C to 140°C
Quantity 5 g
Beilstein 1940558
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 262.05
Percent Purity 97%
Chemical Name or Material 4-Iodophenylacetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only