missing translation for 'onlineSavingsMsg'
Learn More

4-Iodophenyl isothiocyanate, 97%

Catalog No. p-7045953
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1258304 2 g
AAL1258309 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1258304 Supplier Thermo Scientific Chemicals Supplier No. L1258304
Only null left

CAS: 2059-76-9 | C7H4INS | 261.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2059-76-9
Molecular Formula C7H4INS
Molecular Weight (g/mol) 261.08
MDL Number MFCD00041091
InChI Key SNLGBRZZFRSXHA-UHFFFAOYSA-N
Synonym 4-iodophenyl isothiocyanate, 4-iodophenylisothiocyanate, 1-iodo-4-isothiocyanato-benzene, benzene, 1-iodo-4-isothiocyanato, 4-ipitc, acmc-20amg6, 4-iodobenzenisothiocyanate, 1-isothiocyanato-4-iodobenzene, #, benzene,1-iodo-4-isothiocyanato
PubChem CID 74938
IUPAC Name 1-iodo-4-isothiocyanatobenzene
SMILES IC1=CC=C(C=C1)N=C=S

Specifications

Melting Point 74°C to 78°C
Odor Pungent
Quantity 2 g
UN Number UN2923
Beilstein 2412143
Sensitivity Light and moisture sensitive
Formula Weight 261.09
Percent Purity 97%
Chemical Name or Material 4-Iodophenyl isothiocyanate
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only