missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Iodophenetole, 97%
CAS: 699-08-1 | C8H9IO | 248.063 g/mol
$142.82 - $557.84
Chemical Identifiers
| CAS | 699-08-1 |
|---|---|
| Molecular Formula | C8H9IO |
| Molecular Weight (g/mol) | 248.063 |
| MDL Number | MFCD00061124 |
| InChI Key | VSIIHWOJPSSIDI-UHFFFAOYSA-N |
| Synonym | 4-iodophenetole, p-iodophenetole, 1-ethoxy-4-iodo-benzene, 4-ethoxyiodobenzene, 4-ethoxy-4-iodobenzene, benzene, 1-ethoxy-4-iodo, 4-iodophenyl ethyl ether, phenetole, p-iodo, p-ethoxyiodobenzene, ethyl 4-iodophenyl ether |
| PubChem CID | 220700 |
| IUPAC Name | 1-ethoxy-4-iodobenzene |
| SMILES | CCOC1=CC=C(C=C1)I |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2164406
|
Thermo Scientific Chemicals
B2164406 |
5 g |
Each for $142.82
|
|
|||||
|
AAB2164414
|
Thermo Scientific Chemicals
B2164414 |
25 g |
Each for $557.84
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 699-08-1 | |
| 248.063 | |
| VSIIHWOJPSSIDI-UHFFFAOYSA-N | |
| 220700 | |
| CCOC1=CC=C(C=C1)I |
| C8H9IO | |
| MFCD00061124 | |
| 4-iodophenetole, p-iodophenetole, 1-ethoxy-4-iodo-benzene, 4-ethoxyiodobenzene, 4-ethoxy-4-iodobenzene, benzene, 1-ethoxy-4-iodo, 4-iodophenyl ethyl ether, phenetole, p-iodo, p-ethoxyiodobenzene, ethyl 4-iodophenyl ether | |
| 1-ethoxy-4-iodobenzene |
Specifications
| 699-08-1 | |
| 133°C to 134°C (19 mmHg) | |
| C8H9IO | |
| MFCD00061124 | |
| 2042067 | |
| 4-iodophenetole, p-iodophenetole, 1-ethoxy-4-iodo-benzene, 4-ethoxyiodobenzene, 4-ethoxy-4-iodobenzene, benzene, 1-ethoxy-4-iodo, 4-iodophenyl ethyl ether, phenetole, p-iodo, p-ethoxyiodobenzene, ethyl 4-iodophenyl ether | |
| CCOC1=CC=C(C=C1)I | |
| 248.063 | |
| 248.06 | |
| 4-Iodophenetole |
| 26°C to 29°C | |
| >110°C (230°F) | |
| 1.597 | |
| 5 g | |
| Light sensitive | |
| VSIIHWOJPSSIDI-UHFFFAOYSA-N | |
| 1-ethoxy-4-iodobenzene | |
| 220700 | |
| 97% |
Safety and Handling
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only