missing translation for 'onlineSavingsMsg'
Learn More

4-Iodophenetole, 97%

Catalog No. AAB2164406
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2164406 5 g
AAB2164414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2164406 Supplier Thermo Scientific Chemicals Supplier No. B2164406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 699-08-1 | C8H9IO | 248.063 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 699-08-1
Molecular Formula C8H9IO
Molecular Weight (g/mol) 248.063
MDL Number MFCD00061124
InChI Key VSIIHWOJPSSIDI-UHFFFAOYSA-N
Synonym 4-iodophenetole, p-iodophenetole, 1-ethoxy-4-iodo-benzene, 4-ethoxyiodobenzene, 4-ethoxy-4-iodobenzene, benzene, 1-ethoxy-4-iodo, 4-iodophenyl ethyl ether, phenetole, p-iodo, p-ethoxyiodobenzene, ethyl 4-iodophenyl ether
PubChem CID 220700
IUPAC Name 1-ethoxy-4-iodobenzene
SMILES CCOC1=CC=C(C=C1)I

Specifications

Melting Point 26°C to 29°C
Boiling Point 133°C to 134°C (19 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.597
Quantity 5 g
Beilstein 2042067
Sensitivity Light sensitive
Formula Weight 248.06
Percent Purity 97%
Chemical Name or Material 4-Iodophenetole
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only