missing translation for 'onlineSavingsMsg'
Learn More

4-(Iodomethyl)tetrahydro-2h-pyran, 97%, Thermo Scientific™

Catalog No. p-4510370
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Catalog No. Quantity Packaging
CC29940DA 1 g Amber Glass Bottle
1 options

Catalog No. CC29940DA

Supplier: Thermo Scientific Chemicals CC29940DA

Only null left

Chemical Identifiers

CAS 101691-94-5
Molecular Formula C6H11IO
Molecular Weight (g/mol) 226.057
InChI Key MQLFSPBSNWUXSO-UHFFFAOYSA-N
Synonym 4-iodomethyl tetrahydro-2h-pyran, 4-iodomethyl oxane, 4-iodomethyl-tetrahydro-pyran, 4-iodomethyl tetrahydropyran, 2h-pyran, tetrahydro-4-iodomethyl, 4-iodomethyl-tetrahydro-2h-pyran, 2h-pyran,tetrahydro-4-iodomethyl, acmc-20a1ce, 4-iodomethyltetrahydropyran, 4-iodomethyl-tetrahydropyran
PubChem CID 2795507
IUPAC Name 4-(iodomethyl)oxane
SMILES C1COCCC1CI

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 226.05
Percent Purity 97%
Chemical Name or Material 4-(Iodomethyl)tetrahydro-2H-pyran
Hazard Category Danger
Hazard Statement GHS P Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.