accessibility menu, dialog, popup

UserName

CAS RN 101691-94-5

IUPAC Name: 4-(iodomethyl)oxane
Synonyms: 4-(iodomethyl)oxane
Molecular Weight (g/mol): 226.06
Molecular Formula: C6H11IO
InChi Key: MQLFSPBSNWUXSO-UHFFFAOYSA-N
SMILES: ICC1CCOCC1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)